Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B4GALNT2 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734208 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4208 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4208 Supplier Proteintech Group Inc Supplier No. 240241AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

This antibody detects the 55-65 kDa B4GALNT2 protein.

Involved in the synthesis of the Sd(a) antigen (Sia-alpha2,3-[GalNAc-beta1,4]Gal-beta1,4-GlcNAc), a carbohydrate determit expressed on erythrocytes, the colonic mucosa and other tissues. Transfers a beta-1,4-linked GalNAc to the galactose residue of an alpha-2,3-sialylated chain.

Specifications

Antigen B4GALNT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene B4GALNT2
Gene Accession No. Q8NHY0
Gene Alias B4GALGT2, B4GALNT2, B4GALT, Cad, GALGT2, SD, Sda
Gene Symbols B4GALNT2
Host Species Rabbit
Immunogen B4GALNT2 Fusion Protein Ag21166
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124872
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.