Learn More
Invitrogen™ B4GALT4 Polyclonal Antibody

Description
Antibody detects endogenous levels of total B4GALT4.
Specifications
Specifications
| Antigen | B4GALT4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | B4GALT4 |
| Gene Accession No. | O60513, Q66HH1, Q9JJ04 |
| Gene Alias | 9130402O08Rik; B4GALT4; b4Gal-T4; B4galt-IV; Beta-1,4-galactosyltransferase 4; beta-1,4-galactosyltransferase IV; beta-1,4-GalTase 4; beta4Gal-T4; beta4GalT-IV; beta-N-acetylglucosaminyl; Beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase; beta-N-acetylglucosaminyl-glycolipid beta-1,4-galactosyltransferase 4; Lactotriaosylceramide beta-1,4-galactosyltransferase; N-acetyllactosamine synthase; Nal synthase; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase 4; UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 4; UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 4; UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 4; UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 4; UNQ552/PRO1109 |
| Gene Symbols | B4GALT4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.