Learn More
Cytek Biosciences Inc cFluor® B532 Anti-Human CD4 (SK3) - 25 Tests
Supplier:  Cytek Biosciences Inc R720038
Encompass_Preferred
The SK3 monoclonal antibody binds to human CD4, a 59-kDa type I transmembrane glycoprotein in the immunoglobulin superfamily. The CD4 molecule is expressed predominantly on thymocytes and a subpopulation of mature T-helper lymphocytes. It is also present on monocytes at low levels1,2 CD4 plays a role in cell-cell interaction by acting as a co-receptor for MHC class II in antigen recognition. CD4 also has been shown to bind glycoprotein 120 on external envelope of HIV3. The antibody was conjugated to a fluorophore and purified by affinity chromatography.
Catalog No. NC2888843
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.