Learn More
Sino Biological Protein Human CD80 100ug
Supplier:  Sino Biological 10698H02H100
Encompass_Preferred
A DNA sequence encoding the human CD80 (NP 005182 1) (Met1-Asn242) was expressed with the Fc region of human IgG1 at the C-terminus
Catalog No. 50-211-4879
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.