Learn More
Medchemexpress LLC HY-P7324 10ug Medchemexpress, B7-H2/ICOSLG, Fc Chimera (HEK293-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP732410UG
Encompass_Preferred
Medchemexpress, HY-P7324 10ug B7-H2/ICOSLG, Fc Chimera (HEK293-expressed), Human CAS: Recombinant Human B7-H2/ICOSLG, Fc Chimera (HEK293-expressed) is a polypeptide chain containing the C-termimal human IgG1 Fc fragment produced in HEK293 cells. ICOSLG is a number of the B7 family of co-stimulatory molecules. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3146
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.