Learn More
Genscript Corporation B7-H3/CD276 hFc Chimera Human
Supplier:  Genscript Corporation Z03881500
Encompass
B7-H3 a member of the B7 family of immunomodulatory molecules is overexpressed in a wide range of solid cancers B7-H3 binds to activated T cells via an as yet unidentified receptor In assays using sub-optimal amount so anti-CD3 stimulation 2Ig B7 H3 enhances T cell proliferation T cell interferon-gamma (IFN-gamma) production and cytotoxic T cells induction
Catalog No. 50-000-56711