Learn More
Invitrogen™ B7-H3 (CD276) Monoclonal Antibody (M045)

Description
Clone M045 mouse monoclonal antibody detects a 100-120 kDa* protein on SDS-PAGE Native or denatured immunoblots of human LNCaP, MCF-7, NCI-H28, A431, and MDA-MB-231 carcinomas. The antibody detects B7-H3 in membranes and cytoplasm in MCF7 cell after immunocytochemical labeling. The antibody works for western blot, immunoprecipitation, immunocytochemistry, and ELISA capture.
Specifications
Specifications
| Antigen | B7-H3 (CD276) |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | M045 |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1 mg/mL BSA, 50% glycerol and 0.05% sodium azide |
| Gene | CD276 |
| Gene Accession No. | Q5ZPR3 |
| Gene Alias | 4Ig-B7-H3; 6030411F23Rik; AU016588; B7 homolog 3; B7h3; B7-H3; B7RP-2; CD276; CD276 antigen; CD276 molecule; CD276 molecule tv1; costimulatory molecule; PSEC0249; sB7 H3; sB7H3; sCD276; soluble B7 H3; soluble B7H3; soluble CD276; UNQ309/PRO352 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.