Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B7-H6 Monoclonal Antibody (JAM1EW), PE, eBioscience™, Invitrogen™

Catalog No. 12652641
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-652-641 25 Tests
12-652-642 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-652-641 Supplier Invitrogen™ Supplier No. 12652641
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: This JAM1EW monoclonal antibody recognizes human B7-H6 (NCR3LG1) protein on the surface of cells. It recognizes protein on live but not on paraformaldehyde-fixed cells. The JAM1EW clone is specific to B7-H6. When tested in flow cytometry against cells transfected with either B7-H4, B7-H5, B7-H6, or B7-H7, clone JAM1EW only stains positively on B7-H6 transfected cells. Applications Reported: This JAM1EW antibody has been reported for use in flow cytometric analysis. Applications Tested: This JAM1EW antibody has been pre-diluted and tested by flow cytometric analysis of HeLa and HL-60 cell line. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser

B7-H6 (NCR3LG1) is a transmembrane endogenous ligand expressed on the surfaces of tumor cells. B7-H6 binds with NKp30 present on NK cells. Binding of B7-H6 to NKp30 causes the ligation of NKp30 subsequently inducing NK cell activation and target cell cytolysis. B7-H6 is not constitutively expressed on normal tissue. Whether it can be expressed endogenously under certain conditions is under investigation. B7-H6 has been detected on circulating pro-inflammatory CD14(+)CD16(+) monocytes in some patients with sepsis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B7-H6
Applications Flow Cytometry
Classification Monoclonal
Clone JAM1EW
Concentration 5 μL/Test
Conjugate PE
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene NCR3LG1
Gene Accession No. Q68D85
Gene Alias B7 homolog 6; B7H6; B7-H6; DKFZp686I21167; DKFZp686O24166; natural cytotoxicity triggering receptor 3 ligand 1; natural killer cell cytotoxicity receptor 3 ligand 1; NCR3LG1; putative Ig-like domain-containing protein DKFZp686O24166/DKFZp686I21167
Gene Symbols NCR3LG1
Host Species Mouse
Immunogen B7-H6 recombinant protein.
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374383
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.