Learn More
ACROBiosystems Recombinant Protein;Human B7-1 / CD80 Protein, Fc Tag (HPLC-verified);HEK293;1MG;B71,CD80,B7,B7-1,B7.1,BB1,CD28LG,CD28LG1,LAB7
Supplier:  ACROBiosystems B71H52591MG
Encompass_Preferred
Recombinant Protein;Human B7-1 / CD80 Protein, Fc Tag (HPLC-verified);1MG;Human B7-1, Fc Tag (B71-H5259) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession # NP_005182.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human B7-1, Fc Tag (HPLC-verified) (Cat. No. B71-H5259) at 2 μg/mL (100 μL/well) can bind Biotinylated Human CTLA-4 (Cat. No. CT4-H82E4) with a linear range of 50-500 ng/mL (Routinely tested).;Immobilized Human B7-1, Fc Tag (HPLC-verified) (Cat. No. B71-H5259) at 2 μg/mL ( 100 μl/well ) can bind Human CTLA-4, Strep Tag (Cat. No. CT4-H5284) with a linear range of 1-20 ng/mL (QC tested).
Catalog No. 50-165-9040
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.