Learn More
Motic™ BA410E Trinocular Microscope with FITC and TRITC Filter Fluorescence Bundle
Catalog No. 01257540 Shop All Motic Group Products
Change view
Click to view available options
Head Style:
Seidentopf Trinocular
Catalog No. Head Style
01-257-540 Seidentopf Trinocular
1 options
Catalog No. 01-257-540 Supplier Motic™ Supplier No. 1100100404312
Add to Cart
Add to Cart
Request a Quote
This new Fluorescence Bundle includes BA410E 3WLED Trinocular Sextuple Microscope (light split 80/20), Complete EPI-Fluorescence Attachment, EGFP/FITC/Cy2/AlexaFluor 488 and TRITC/Cy3/TagRFP/AlexaFluor 546.
  • Display maximum functionality and ergonomics in all its mechanical aspects
  • Elegant and robust, the microscope stand follows all needs for an intense daily use
  • Eyepiece tubes guarantee a fatigue-free work in terms of viewing angle and improved interpupillary distance, while the 'butterfly' movement may increase the viewing height by 60 mm for individual positioning
  • Diopter Adjustment
  • Koehler Quartz halogen 12 V/50 W with intensity control
  • CCIS™ Infinity concept presents this apochromatic level of performance in a complete set of optional bright field objectives, compatible to all BA series models of upright microscopes
  • Highest quality demands from pathology, histology and hematology can now be satisfied for a reliable diagnosis in routine and research work of hospitals and other biomedical labs
  • Based on an encoded 6-fold nosepiece which memorizes the light intensity setting for each nosepiece position
  • System replicates this intensity setting once the respective position is swung in again
  • Any new setting is taken as an overwriting of the former setup
Create a digital environment with Moticam: digitize your lab with our Moticam solutions. Look at your samples in your computer, your tablet or smartphone, a TV screen or a projector and view, capture, edit, measure, make reports or share your work with ease with anyone in just a few clicks. Motic™'s software solutions included in the packages show a user-friendly interface, making data sharing possible in a collaborative environment, encouraging teamwork and collective learning experiences.
Specifications
Eyepiece N-WF 10X/22 mm, with diopter adjustment
Head Style Seidentopf Trinocular
Lighting Type Quartz Halogen
Magnification Power 4X, 10X, 40XS, 100XS (oil)
Model BA410E
Objective Types Infinity Corrected Plan Achromatic
Product Type Trinocular Microscope
Nosepiece Reversed Sextuple
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov