Learn More
Description
PA1-754 detects human and mouse beta-secretase 2 (BACE2) from transfected cells. This product has recently been remanufactured in new rabbits as Cat. # PA1-1754. PA1-754 has been successfully used in Western blot and ICC/IF procedures. By Western blot this antibody detects an ~58 kDa protein representing BACE2 from SY-SY5Y cells overexpressing the human gene. The PA1-754 immunogen is amino acid residues 503-518 from human BACE2. This sequence is completely conserved in mouse. The PA1-754 immunizing peptide (Cat. # PEP-167) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | BACE2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1 mg/mL BSA and 0.05% sodium azide |
| Gene | BACE2 |
| Gene Accession No. | Q9JL18, Q9Y5Z0 |
| Gene Alias | 1110059C24Rik; 56 kDa aspartic-like protease; AEPLC; AI850424; ALP56; ARP1; asp 1; ASP1; ASP21; Aspartic-like protease 56 kDa; Aspartyl protease 1; Bace2; BAE2; be; beta secretase 2; Beta-secretase 2; beta-site amyloid beta A4 precursor protein-cleaving enzyme 2; beta-site amyloid precursor protein cleaving enzyme 2; Beta-site APP cleaving enzyme 2; beta-site APP-cleaving enzyme 2; CDA13; CEAP1; Down region aspartic protease; Down syndrome region aspartic protease; DRAP; memapsin 1; memapsin-1; Membrane-associated aspartic protease 1; theta-secretase; transmembrane aspartic proteinase Asp1; UNQ418/PRO852 |
| Gene Symbols | BACE2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
