Learn More
Invitrogen™ BACH1 Polyclonal Antibody

Description
PA1-9537 detects BACH1 in human samples. PA1-9537 has been successfully used in Western blot procedures. By Western blot, PA1-9537 detects a 100 kDa protein representing BACH1. PA1-9537 immunogen is a purified recombinant BACH1.3 corresponding to the chromosome 21 gene (Human).
Specifications
Specifications
| Antigen | BACH1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | BACH1 |
| Gene Accession No. | O14867 |
| Gene Alias | 6230421P05Rik; AI323795; BACH1; BACH-1; basic region leucine zipper transcriptional regulator BACH1; BTB and CNC homolog 1; BTB and CNC homology 1; BTB and CNC homology 1 transcription factor; BTB and CNC homology 1, basic leucine zipper transcription factor 1; BTB domain and CNC homolog 1; BTBD24; HA2303; Transcription regulator protein BACH1 |
| Gene Symbols | BACH1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.