Learn More
Sigma Aldrich Fine Chemicals Biosciences BachiR Rotavapor R 100 co
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7419821SET
Encompass
The rotary evaporator R-100 is a high-quality entry level Rotavapor to meet the essential needs in classical laboratory applications. It is capable of a manually adjustable rotation speed of 20 to 280 rpm and equipped with an action jack lift for manual lifting of the evaporating flask. Included is the selected condenser 1 L receiving and evaporating flasks and a vapor duct. All accessories are included for out of the box usage.
Catalog No. 50-290-8577
Only null left
Add to Cart
Request a Quote