Learn More
CROSSTEX INTERNATIONAL Biological Indicators - ISO 11138-4 Compliant, 100/pk
Supplier:  CROSSTEX INTERNATIONAL BG106DH
Encompass_Preferred
Crosstex offers a biological indicator for Dry Heat sterilization which is compliant with ISO 11138-4.
Catalog No. NC1041789
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.