Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bacitracin, Fisher BioReagents

Catalog No. BP29501 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP29501 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP29501

missing translation for 'validMainframeMsg'

Inhibitor of protein disulfide isomerase. Interferes with building blocks of peptidoglycan bacterial cell wall

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-87-4
Molecular Formula C66H103N17O16S

Specifications

Type Bacitracin
Physical Form Powder/Solid
pH 6.0 to 7.0
Melting Point 221°C
Source Bacillus licheniformis
Loss on Drying < 5.0%
Packaging Amber Glass
Potency > 70 U/mg
Color Beige
Quantity 1 g
Merck Index 15, 927
Solubility Information Soluble in water
Formula Weight 1422.69
Chemical Name or Material Bacitracin
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
May cause an allergic skin reaction. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.