accessibility menu, dialog, popup

UserName

Viewing profile as user:
ZnH3CH3CNH2NSOHNCH3H3CONHHOOOHNH3CH3CONHNHONH2OHNOOHONHONNNHONHOHNOH2NNHOCH3CH3

CAS RN 1405-87-4

IUPAC Name: 4-(2-{[2-(1-amino-2-methylbutyl)-4,5-dihydro-1,3-thiazol-4-yl]formamido}-4-methylpentanamido)-4-[(1-{[18-(3-aminopropyl)-12-benzyl-15-(butan-2-yl)-3-(carbamoylmethyl)-6-(carboxymethyl)-9-[(4H-imidazol-4-yl)methyl]-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptaazacyclopentacosan-21-yl]carbamoyl}-2-methylbutyl)carbamoyl]butanoic acid zinc
Synonyms: bacitracin
Molecular Weight (g/mol): 1488.09
Molecular Formula: C66H103N17O16SZn
InChi Key: QSNOBVJFKSQBBD-UHFFFAOYNA-N
SMILES: [Zn].CCC(C)C(N)C1=NC(CS1)C(=O)NC(CC(C)C)C(=O)NC(CCC(O)=O)C(=O)NC(C(C)CC)C(=O)NC1CCCCNC(=O)C(CC(N)=O)NC(=O)C(CC(O)=O)NC(=O)C(CC2C=NC=N2)NC(=O)C(CC2=CC=CC=C2)NC(=O)C(NC(=O)C(CCCN)NC1=O)C(C)CC
Physical Form 
  • (1)
  • (7)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
Type 
  • (3)
  • (6)
Color 
  • (1)
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (3)
  • (1)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (6)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)