Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Bacitracin, potency min. 60 Units/mg

Catalog No. AC226100050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226100050 5 g Glass Bottle
AC226100250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC226100050 Supplier Thermo Scientific Chemicals Supplier No. 226100050
Add to Cart
This item is not returnable. View return policy

Bacitracin, CAS # 1405-87-4, is cyclic polypeptide-based antibiotic which exhibits activity against microbes causing pneumonia, skin, and eye infections.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Bacitracin is a mixture of at least nine different cyclic polypeptides, originally isolated from Bacillus bacteria in 1945

• Bacitracins work by binding divalent metal cations and preventing the hydrolysis of lipids into usable cell wall components

Applications

• Bacitracin has been paired with nanoparticles in some laboratory studies to increase its effectivity

• It is suitable for the laboratory study of bacterial resistance, infection, and cross-reactivity

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-87-4
Molecular Formula C66H103N17O16S
MDL Number MFCD00062640

Specifications

Type Bacitracin
Physical Form Powder
Melting Point 221.0°C to 225.0°C
Infrared Spectrum Authentic
Loss on Drying 5.0% max. (1 g, 60°C, 3 Hrs) (P2O5) (not exceeding 0.1 kPa)
Packaging Glass Bottle
Color Beige or White
Concentration ≥60 I.U./mg (potency, on dried subst.)
Quantity 5 g
Merck Index 15, 927
Solubility Information Solubility in water: soluble. Other solubilities: soluble in glacial acetic acid, methanol and, alcohol, practically insoluble in ether, chloroform and, acetone
Formula Weight 1422.71
Percent Purity 60 I.U./mg min. (potency, on dried subst.)
Chemical Name or Material Bacitracin
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 215-786-2
RTECSNumber CP0175000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.