Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bacitracin

Catalog No. AAJ6243206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6243206 5 g
AAJ6243214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6243206 Supplier Thermo Scientific Chemicals Supplier No. J6243206
  • $107.40
    $91.65 / Each of 1

    Save $15.75

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

Bacitracin, CAS # 1405-87-4, is cyclic polypeptide-based antibiotic which exhibits activity against microbes causing pneumonia, skin, and eye infections. | CAS: 1405-87-4 | C66H103N17O16S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Bacitracin is a mixture of at least nine different cyclic polypeptides, originally isolated from Bacillus bacteria in 1945
  • Bacitracins work by binding divalent metal cations and preventing the hydrolysis of lipids into usable cell wall components

Applications

  • Bacitracin has been paired with nanoparticles in some laboratory studies to increase its effectivity
  • It is suitable for the laboratory study of bacterial resistance, infection, and cross-reactivity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-87-4
Molecular Formula C66H103N17O16S
MDL Number MFCD00062640

Specifications

Type Bacitracin
Physical Form Powder
Melting Point 222°C
Color White
Quantity 5 g
Merck Index 14,934
Solubility Information Soluble in water.
Formula Weight 1421.6
Chemical Name or Material Bacitracin
Hazard Category H317-H336-H373
Precautionary Statement P260-P271-P272-P280g-P302+P352-P304+P340-P312-P314-P333+P313-P363-P501c
EINECSNumber 215-786-2
RTECSNumber CP0175000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.