Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bacitracin, USP Grade, G-Biosciences
SCI_ED

Catalog No. S162510
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S162510 50 g
S139088 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S162510 Supplier G-Biosciences Supplier No. RC897
Add to Cart
Add to Cart

Bacitracin is a cyclic polypeptide antibiotic isolated from Bacillus subtilis.

Bacitracin is a cyclic polypeptide antibiotic isolated from Bacillus subtilis. It is made up of a variety of antimicrobial peptide fractions including A, A1, B, B1, B2, C, D, E, F, G, and X. This standard grade product is a mixture of these fractions, with bacitracin A exhibiting the most potent antimicrobial activity. Bacitracins B1 and B2 exhibit about 90% of the antimicrobial activity of bacitracin A when together.

Bacitracin targets the cell wall of gram-positive organisms such as Streptococcus pyogenes and Staphylococcus aureus. Bacitracin also functions as a useful tool to distinguish between group A and group B Beta-hemolytic Streptococcus species, which are susceptible and resistant, respectively.

Chemical Identifiers

CAS 1405-87-4
Molecular Weight (g/mol) 1422.69 g/mol

Specifications

Technique HPLC, Spectrometry, UV/Vis, Ion Chromatography, Mass Spectrometry
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, NIST
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 50 g
Percent Purity 98%
Grade USP
Chemical Name or Material Bacitracin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.