Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bacitracin USP Micronized, Medisca™
GREENER_CHOICESDP

Catalog No. 18048081 Shop All Medisca Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-048-081 10 g
18-048-106 25 g
18-048-130 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-048-081 Supplier Medisca Supplier No. 001501
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1405-87-4

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Color White to Off-White
Quantity 10 g
Percent Purity ≥ 65 Bacitracin units/mg
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.