Learn More
Selleck Chemical LLC Baclofen 25mg 1134-47-0
SDP
Supplier:  Selleck Chemical LLC S484025MG
Encompass_Preferred
Baclofen is a gamma-amino-butyric acid (GABA) derivative used as a skeletal muscle relaxant and central nervous system depressant. It activates GABA receptors, specifically the GABAB receptors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8442
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.