Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BacMam Akt [pSer473] Cellular Assay Kit

Catalog No. A12899
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12899 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A12899 Supplier Thermo Scientific™ Supplier No. A12899

The combination of baculovirus-mediated gene delivery (BacMam) with LanthaScreen™ Cellular Assay technology enables a platform for the analysis of specific posttranslational modifications of various substrates including AKT.

The combination of baculovirus-mediated gene delivery (BacMam) with LanthaScreen™ Cellular Assay technology enables a platform for the analysis of specific posttranslational modifications of various substrates including AKT. BacMam provides a convenient genetic delivery tool for a GFP-AKT fusion protein in the cell line of interest. This kit describes an HTS-compatible cellular immunoassay measuring phosphorylation of GFP-AKT at Ser473.

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Cell-based terbium modification activity (transient), Cell-based terbium phospho-specific activity (transient)
Cell State Cells Not Included
Content And Storage Kit contains Terbium-labeled anti-AKT antibody, BacMam AKT Virus, and 6X Lysis Buffer. Store as indicated in handling documentation.
Detection Method Fluorescence
For Use With (Application) TR-FRET
Human Protein Class Kinases, Cell Signalling
Product Line LanthaScreen
Quantity 1 kit
Readout End Point
Reporter Gene GFP (EmGFP)
Target Entry AKT1
Type Cellular Assay Kit
System Type LanthaScreen™
Druggable Target Kinases
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.