Learn More
Thermo Scientific™ BacMam Akt [pThr308] Cellular Assay Kit
Description
The combination of baculovirus-mediated gene delivery (BacMam) with LanthaScreen™ Cellular Assay technology enables a platform for the analysis of specific posttranslational modifications of various substrates including AKT. BacMam provides a convenient genetic delivery tool for a GFP-AKT fusion protein in the cell line of interest. This kit describes an HTS-compatible cellular immunoassay measuring phosphorylation of GFP-AKT at Thr308.
Specifications
Specifications
| Assay Entry | Cell-based terbium modification activity (transient), Cell-based terbium phospho-specific activity (transient) |
| Cell State | Cells Not Included |
| Content And Storage | Kit contains Terbium-labeled anti-AKT antibody, BacMam AKT Virus, and 6X Lysis Buffer. Store as indicated in handling documentation. |
| Detection Method | Fluorescence |
| For Use With (Application) | TR-FRET |
| Human Protein Class | Kinases, Cell Signalling |
| Product Line | LanthaScreen |
| Quantity | 1 kit |
| Readout | End Point |
| Reporter Gene | GFP (EmGFP) |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.