Learn More
Invitrogen™ BacMam ERK2 [pThr185/pTyr187] Cellular Assay Kit
Description
The BacMam ERK2 [pThr185/pTyr187] Cellular Assay Kit is a high-throughput screening (HTS) compatible cellular immunoassay for the measuring of phosphorylation of GFP-ERK2. The BacMam ERK2 Cellular Assay Kit can be used with numerous transformed cell lines, primary cells, and stem cells, and allows for the interrogation of ERK2, either as a kinase or GPCR readout.
- Choose cell types that are relevant to your needs
- Get results in weeks instead of months
- Perform Assays on Your Schedule
Use Cell Types Relevant to Your Needs
Understanding signaling in physiologically relevant cell backgrounds is extremely important when predicting the real effects of compounds in humans prior to clinical trials. With the BacMam ERK2 [pThr185/pTyr187] Cellular Assay Kit you can choose the appropriate cell type for your research needs, enabling you to design your own experiments, and to reach your research goals faster.
BacMam ERK2 Reagent Compatible Cells
The BacMam reagent included in the kit has been used to successfully transduce numerous transformed cell lines (e.g. U2-OS, HEK 293, and HeLa), primary cells (e.g., fibroblasts, hepatocytes, cardiovascular cells and epithelial cells), and stem cells (e.g. neuronal and mesenchymal) at high transduction efficiency and low levels of cytotoxicity. However, cells of hematopoietic origin show consistently poor transduction by BacMam technology, and are not recommended for use with the current technology.
Develop Assays Faster
The BacMam platform enables transient target expression allowing for reduced time to an HTS ready assay versus traditional stable cell line creation. Traditional stable cell line development requires months of culturing, optimization, and cloning of your cell line. This requires considerable time and resources. With BacMam ERK2 a user can get results in weeks versus months.
Cells Ready When You Are
BacMam Erk2 Reagent is ideal to transduce large quantities of cells in batch mode; transduced cells can be stored frozen in aliquots for later use. This saves you the delays associated with culturing cells. The cells are ready whenever you are. Cells can be assayed typically within hours of thawing, providing assay-ready cells when you need them, with no loss of activity.
Specifications
Specifications
| Assay Entry | Cell-based terbium modification activity (transient) |
| Content And Storage | Kit contains Terbium-labeled anti-ERK2 [pThr185/pTyr187] antibody, BacMam ERK2 Virus, 6X Lysis Buffer, and BacMam Enhancer Solution. Store as indicated in handling documentation. Cells not included |
| Delivery Type | Baculovirus (BacMam System) |
| Detection Method | Fluorescence |
| For Use With (Application) | TR-FRET |
| Gene Symbol | ERK2 |
| Quantity | 1 Kit |
| Readout | End Point |
| Target Entry | ERK2 |
| Type | Cellular Assay Kit |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.