Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BacMam LRRK2 Reagent

Catalog No. A13388
Encompass_Preferred
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13388 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A13388 Supplier Invitrogen™ Supplier No. A13388

BacMam LRRK2 Reagent

BacMam LRRK2 Wild Type Reagent is a modified insect cell virus (baculovirus) that enables easy, efficient, and titratable overexpression of full-length wild-type LRRK2 in your choice of cellular background, including primary neurons and neuroblastoma cell lines.

BacMam LRRK2 Wild Type Reagent Lets You:

  • Easily overexpress full-length LRRK2 in most cellular backgrounds
  • Conduct the LRRK2 cellular assays you need
  • Understand the effects of therapeutically relevant LRRK2 mutations

    Examine LRRK2 Mutations with BacMam Reagents
    Use therapeutic mutant reagents G2019S (Cat. no. A13389) and D1994A (Cat. no. A13390) along with the wild-type LRRK2 reagent to enable your research into LRRK2 mutations, including exploration of differences in cellular response to stimulus among different mutations.

    Expand Your Battery of Cellular Assays for LRRK2
    Once full length LRRK2 has been introduced into your cellular background, the transduced cells allow you the flexibility to conduct many types of LRRK2 experiments, including studies of regulation of physiologically functional pathways (including signaling pathways leading to function), and identification of potential physiological substrates for LRRK2.

    Overexpress Full-Length LRRK2 in Most Mammalian Cell Backgrounds
    BacMam technology utilizes a modified baculovirus to deliver and express genes in mammalian cells with minimal effort and toxicity. It enables efficient, titratable LRRK2 protein expression in various mammalian cellular backgrounds, including cell lines that may not be compatible with traditional transfection methods, such as primary neurons and neuroblastoma cell lines.

    For research use only. Not intended for any human or animal therapeutic or diagnostic use.

    Related Links:
  • LRRK2 tools for advancing Parkinson's disease research
  • Kinase protein portfolio
  • LanthaScreen™ Eu Kinase Binding Assay
  • LanthaScreen™ Activity Assay
  • Learn More About BacMam Technology
  • TRUSTED_SUSTAINABILITY

    Specifications

    Formulation Recombinant baculovirus particles suspended in Gibco™ Sf-900™ III SFM media supplemented with 3% heat-inactivated fetal bovine serum (hiFBS)
    Type BacMam LRRK2 Reagent
    Target Kinases
    Delivery Type Baculovirus (BacMam System)
    Quantity 15 mL
    Content And Storage Recombinant baculovirus particles suspended in Gibco™ Sf-900™ III SFM media (Cat# 12658) supplemented with 3% heat-inactivated fetal bovine serum (hiFBS) (Cat# 10100). Store at 4°C, keep sterile, protect from light, DO NOT freeze, aliquot upon arrival.
    Gene Symbol LRRK2
    Human Protein Class Kinases
    Shipping Condition Wet Ice
    For Use With (Application) Overexpression of full-length wild-type LRRK2

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.