Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva bacteria genomicPrep Mini Spin Kit

Catalog No. 28904258 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
250 Preps
50 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
28-904-258 50 Preps
45-001-494 250 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 28-904-258 Supplier Cytiva Supplier No. 28904258
Add to Cart
Add to Cart

Includes

illustra bacteria genomicPrep Mini spin columns prepacked with novel silica membrane; Lyophilized proteinase K powder; Lysis solutions, wash and elution buffers; 2mL Microcentrifuge collection tubes; a full booklet with a detachable, quick reference protocol card

Designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from Gram-negative (G-) and Gram-positive (G+) bacteria

The illustra Bacteria genomicPrep Mini Spin Kit can be used with a wide range of bacterial strains such as DH5a, TOP10, JM109, Bacillus subtilis etc. The kit produces highly pure and intact genomic DNA with OD260/280 values of 1.75-1.85. Use different elution volumes to isolate genomic DNA at concentrations suitable for most downstream applications including restriction enzyme digestion, RT-PCR, and long PCR.

The method uses a lysis solution in combination with proteinase K to gently release genomic DNA into solution from bacteria cells. The DNA is deproteinated in the extraction solution and the genomic DNA is then bound onto a silica column in the presence of chaotropic solution. Contaminants are removed during wash steps and the purifed DNA is eluted with prewarmed TE solution.

  • Total DNA extraction time under one hour with reduced lysis time
  • Can be used to purify 4 to 8μg of genomic DNA from 2 × 109 bacteria cells with input amounts ranging from 1 × 109 to 4 × 109 bacteria cells
  • Spin columns prepacked with a silica membrane—provide excellent reproducibility and consistency
  • Validated in several downstream applications including RT PCR, long PCR, and restriction digest
  • Color-coded caps for ease of use

TRUSTED_SUSTAINABILITY

Specifications

Description illustra bacteria genomicPrep Mini Spin Kit is designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from Gram-negative (G-) and Gram-positive (G+) bacteria.
For Use With (Application) For the rapid extraction and purification of high-quality genomic DNA from gram-positive and gram-negative bacteriaPurified gDNA is suitable for molecular biology applications including cloning, restriction enzyme digestion, PCR and genotyping
Includes 2 x 30mg Proteinase K, 15mL Lysis Buffer type 2, 2 x 1.5mL Lysis Buffer type 3, 2 x 165mL Lysis Buffer type 4, 30mL Wash Buffer type 6, 60mL Elution Buffer type 5, 5 x 50 Illustra™ bacteria mini columns, 5 x 50 collection tubes
No. of Reactions 50 Preps
Product Type illustra bacteria genomicPrep Mini Spin Kit
Purity 1.7
Sample Volume 2 x 109 cells [A600 = 2.0]
Test Time 40 min [Gram-negaitive Bacteria], 55 min [Gram-positive Bacteria]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.