Learn More
Bacterial Acid Methyl Ester (BAME) Mix solution (10 mg/mL total concentration in methyl caproate),, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Packaging | Ampule |
| CAS | 106-70-7 |
| Flash Point | 43°C |
| Physical Form | Solution |
| Type | Analytical Standard |
| Quantity | 1 mL |
| Grade | Analytical Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.