Learn More
Cytiva (Formerly Pall Lab) Bacterial Air Vents - 1 m (nominal), 37 mm, sterile (10/pkg)
Supplier:  Cytiva 4308N
Encompass_Preferred
For use as vents for receiving vessels or as sterile breathing barriers.-Vents for receiving vessels-Sterile breathing barrier-AutoclavableUse in-line for sterile air/gas delivery to culture vessels, bioisolation of vacuum source, flushing instruments, or cleaning parts.Can be sterilized by autoclaving. Polypropylene housing. With 1m hydrophobic glass laminate filter. Connector is 6.49.5mm (1/43/8") stepped hose barb; inner taper accepts standard male slip Luer. Maximum pressure 5.2bar (75psi). Effective filtration area: 7.5cm2. Dimensions: 4.5dia.x5.3Lcm (125/32x23/32"). This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3033
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.