Learn More
Promega Bacterial Strain NM522, Glycerol Stock
Catalog No. PRP2301 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
500 μL
Catalog No. Quantity
PRP2301 500 μL
1 options
Catalog No. PRP2301 Supplier Promega Supplier No. P2301
Add to Cart
Add to Cart
NM522 contains an F' episome, which is required for production of ssDNA and for blue/white color selection.
  • Produces ssDNA with F´ Episome
  • Can be used for blue/white color selection when screening colonies
Specifications
Content And Storage <-65°C
Quantity 500 μL