Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bacteriological Agar, Ultrapure

Catalog No. AAJ10906P2
Change view
Click to view available options
Quantity:
1 lb.
5 lb.
25 lb.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ10906P2 1 lb.
AAJ10906P5 5 lb.
AAJ10906Q1 25 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ10906P2 Supplier Thermo Scientific Supplier No. J10906P2
Add to Cart

Used as a gelling agent in preparation of bacteriological culture media.

Used extensively as a gelling agent in the preparation of bacteriological culture media. Its main attributes are superior transparency, very reliable reproducibility and absence of inhibitors capable of hindering optimal development of microorganisms. Extraneous matter reduced to a minimum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Superior transparency
  • Reliable reproducibility
  • Absence of inhibitors capable of hindering optimal development of microorganisms
  • Extraneous matter reduced to minimum
  • Format: White to light cream-colored powder
  • CAS: 9002-18-0
  • MDL Number: MFCD00081288
TRUSTED_SUSTAINABILITY

Specifications

Color White to light cream-colored
Form Powder
Grade Ultrapure
Media Type Bacteriological Agar
Product Type Nutrient Agar
For Use With (Application) Used as a Gelling Agent in Preparation of Bacteriological Culture Media.
Packaging Type Plastic Bottle
Quantity 1 lb.
EINECSNumber 232-658-1
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.