Learn More
Description
Used extensively as a gelling agent in the preparation of bacteriological culture media. Its main attributes are superior transparency, very reliable reproducibility and absence of inhibitors capable of hindering optimal development of microorganisms. Extraneous matter reduced to a minimum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Superior transparency
- Reliable reproducibility
- Absence of inhibitors capable of hindering optimal development of microorganisms
- Extraneous matter reduced to minimum
- Format: White to light cream-colored powder
- CAS: 9002-18-0
- MDL Number: MFCD00081288
Specifications
Specifications
| Color | White to light cream-colored |
| Form | Powder |
| Grade | Ultrapure |
| Media Type | Bacteriological Agar |
| Product Type | Nutrient Agar |
| For Use With (Application) | Used as a Gelling Agent in Preparation of Bacteriological Culture Media. |
| Packaging Type | Plastic Bottle |
| Quantity | 5 lb. |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.