Learn More
Thermo Scientific™ BactiSwab™ Gel Collection and Transport Swabs: Dual Swab Amies Clear
Catalog No. R723010
Change view
Click to view available options
Packaging:
50/Pk.
500/Pk.
Swab Configuration:
Dual Applicator
Single Applicator
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Swab Configuration
R723010 500/Pk. Dual Applicator
R723005 50/Pk. Single Applicator
R723000 500/Pk. Single Applicator
R723015 50/Pk. Dual Applicator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. R723010 Supplier Thermo Scientific™ Supplier No. R723010
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel BactiSwab Gel Amies Clear are supplied with double plastic shaft.
Thermo Scientific™ Remel BactiSwab Gel Amies Clear are supplied with double plastic shaft.
Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing gel transport medium with or without charcoal.
  • Blue cap
  • Round-bottom tube for easy racking and seamless molding to ensure strength and integrity
  • Exclusive zip-seal packaging to promote optimum media performance
  • Color-coded cap with protective overskirt
  • Inert, non-toxic rayon swab tip for good sample retrieval and absorption
TRUSTED_SUSTAINABILITY
Specifications
Format Tube
Media Type Amies Clear
Swab Configuration Dual Applicator
Closure Color Blue
Description Amies Clear
Packaging 500/Pk.
Quantity 1 Tube/Envelope
Shaft Material Plastic