Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BactiSwab™: Wire Shaft Liquid Stuart

Catalog No. R723130
Change view
Click to view available options
Packaging:
100/Pk.
50/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
R723130 100/Pk.
R723135 50/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R723130 Supplier Thermo Scientific™ Supplier No. R723130
Add to Cart
Add to Cart
This item is not returnable. View return policy

Supplied with a mini-tip swab suitable for ear, nose, throat, eye, male urethral and pediatric specimen collection.

Thermo Scientific™ Remel BactiSwab: Wire Shaft Liquid Stuart is supplied with a mini-tip swab suitable for ear, nose, throat, eye, male urethral and pediatric specimen collection.

Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing a sponge impregnated with liquid transport medium.
  • Straight aluminum shaft
  • Green cap
TRUSTED_SUSTAINABILITY

Specifications

Format Tube
Media Type Liquid Stuart
Closure Color Green
Description Liquid Stuarts
For Use With (Application) Healthcare
Packaging 100/Pk.
Shaft Material Wire
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.