Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Bacto™ Brain Heart Infusion

Catalog No. DF0037070 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF0037-07-0 2 kg Pail
DF0037-17-8 500 g Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DF0037-07-0 Supplier BD Difco™ Supplier No. 237200
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in the cultivation of fastidious and nonfastidious microorganisms. BD Difco™ Bacto™ Brain Heart Infusion is a general-purpose medium used to cultivate aerobic and anaerobic bacteria from a variety of clinical and nonclinical materials.

BHI broth is used for the cultivation of a wide variety of microorganisms, including bacteria (streptococci, pneumococci and meningococci), yeasts and molds. It serves as a base for supplemented media containing 0.1% agar, Fildes enrichment or 6.5% sodium chloride.

  • To use dehydrated product: suspend 37g in 1L of purified water
  • Displays a good cultural response to Neisseria meningitides 13090, Streptococcus pneumonia 6305 and Streptococcus pyogenes 19615
  • pH (3.7% solution, 25°C): 7.4 ± 0.2

Certifications

AOAC, BAM, CCAM, CLSI, CMPH2, COMPF, EPA, ISO, MCM9, SMD, SMWW, USDA

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, CCAM, CLSI, CMPH2, COMPF, EPA, ISO, MCM9, SMD, SMWW, USDA
Color Light tan
Content And Storage 2 to 25°C
Form Dehydrated (free-flowing, homogenous)
Media Type Brain Heart Infusion
Product Type Nutrient Agar
Formulation Calf brain (infusion from 200g) 7.7g/L, Beef heart (infusion from 250g) 9.8g/L, Proteose Peptone 10g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms, including aerobic and anaerobic bateria, yeasts and molds
Packaging Type Pail
Quantity 2 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.