Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Bacto™ Tryptose Phosphate Broth
GREENER_CHOICE

Catalog No. b60200
Change view
Click to view available options
Quantity:
10 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B60200 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B60200 Supplier Gibco™ Supplier No. 260200
Only null left
Add to Cart
Add to Cart

Bacto Tryptose Phosphate Broth is an infusion-free buffered medium recommended for the cultivation of fastidious, pathogenic microorganisms.

Bacto Tryptose Phosphate Broth is an infusion-free buffered medium recommended for the cultivation of fastidious, pathogenic microorganisms. It is valuable in tissue culture procedures where the peptone content is considered to be a stimulating factor for cells.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2°C to 25°C in cool, dry conditions in well-sealed containers.
Form Dehydrated
Product Type Bacterial Growth Medium
Product Line Bacto
Quantity 10 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.