Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Bacto™ CD Supreme Feed (2X), powder

Catalog No. A9378301
Change view
Click to view available options
Quantity:
250 g
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A9378301 250 g
A9378302 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A9378301 Supplier Gibco™ Supplier No. A9378301
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemically defined nutrient feed for optimized E. coli plasmid DNA and protein production.

Gibco Bacto CD Supreme Feed (2X) is an animal origin–free, chemically defined microbial feed specifically formulated to extend culture performance and increase productivity in Escherichia coli (E. coli) fed-batch processes. Designed to deliver a balanced nutrient supply, this hydrolysate-free formulation supports robust cell growth, higher biomass accumulation, and improved product yield across a broad range of E. coli expression strains.

Features of Bacto CD Supreme Feed include:

  • Increased productivity with 10-fold higher plasmid DNA production compared to batch mode
  • Consistency with a 100% chemically defined, hydrolysate-free composition that minimizes variability
  • Streamlined operations using a one-part, ready-to-use formulation that eliminates the need for complex feed preparation
  • Easy integration with Bacto CD Supreme FPM and standard sterilization methods for scalable implementation
  • Simplified compliance with an animal origin–free formulation that supports global regulatory requirements

Developed for compatibility with both Bacto CD Supreme Fermentation Production Medium (FPM) Plus (Cat. Nos. A9378401, A9378501, A9378502) and Bacto CD Supreme FPM (Cat. Nos. A4973701, A4973702), Bacto CD Supreme Feed provides sustained nutrient delivery during fed-batch fermentation that increases biomass accumulation, extends bioreactor runs, and enhances plasmid DNA and protein yield.

As a chemically defined formulation, Bacto CD Supreme Feed eliminates the performance variability associated with peptone-based feeds and supports consistent outcomes across diverse E. coli strains used in plasmid DNA and recombinant protein production.

Offered in convenient liquid and dry powder (DPM) formats, Bacto CD Supreme Feed ensures ease of use, scalability, and reliability from development to large-scale manufacturing.

Specifications

Content And Storage Store in the dark at +2°C to +8°C
Form Dehydrated Powder
Manufacturing Origin United States
Product Type E. coli Feed
Sterility Non-sterile
Target Organism Class E. coli
Test Type Microbial Cultivation
Formulation Animal Origin-free, Chemically-defined
For Use With (Application) Microbial Fermentation
Product Line Bacto CD Supreme
Quantity 250 g
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.