Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Bacto™ Dehydrated Culture Media: Heart Infusion Broth

Catalog No. DF0038079 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
2 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0038-07-9 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0038-07-9 Supplier BD Supplier No. 238100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Isolation and cultivation of wide variety of fastidious microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BAM, CCAM, COMPF, EP
Content And Storage Refrigerate
Form Dehydrated
Media Type Heart Infusion Broth
Product Type Microbiology Media
Formulation Beef heart infusion from 500g 10g/L, Tryptose 10g/L, Sodium Chloride 5g/L
For Use With (Application) Isolation and cultivation of a wide variety of fastidious microorganisms
Packaging Type Pail
Quantity 2 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.