Learn More
Gibco™ Bacto™ Neopeptone

Description
- Bacto Neopeptone is an animal origin (AO) enzymatic digest of bovine (milk) and porcine protein
- This nutritional supplement provides vitamins, nucleotides, minerals, and peptides in a variety of sizes to any culture formulation
- It has been used as a component of culture media in the cultivation of human pathogens, notably Bordetella pertussis and group A streptococci
- Also used with Borrelia burgdorferi and Clostridium species, to produce vaccines for animal health application
- Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Applications of Bacto Neopeptone include:
- Recommended for use in media for detection of fungi
- Reported to provide nutrients for support of spirochetes and protozoa
- Microbial fermentation
- Production of vaccines for animal health applications
Specifications
Specifications
| Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| For Use With (Application) | Production of vaccines for animal health applications |
| Product Line | Bacto |
| Quantity | 10 kg |
| Source | Animal origin, bovine and porcine |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.