Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Bacto™ Proteose Peptone No. 4
GREENER_CHOICE

Catalog No. 211715TF Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-171-5TF 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-171-5TF Supplier Gibco™ Supplier No. 211715
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

A member of the Gibco Bacto Proteose Peptones, which are enzymatic digests of protein.

A spray-dried version of Bacto™ Proteose Peptone. It offers the same beneficial nutrients as Bacto™ Proteose Peptone for growth promotion and toxin production in a wide range of fastidious microorganisms.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Dehydrated Peptone
For Use With (Application) Microbiological culture media and in producing bacterial toxins
Product Line Bacto
Quantity 10 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.