Learn More
Gibco™ Bacto™ Soytone

Description
- An enzymatic digest of soy that uses an animal origin (AO) (porcine) enzyme for digestion
- The naturally high concentrations of vitamins and carbohydrates found in soy make Bacto Soytone a nutritious carbohydrate source for those not concerned about the processing step involving an animal-based material
- This supplement offers a unique nutritional profile as compared to other soy products
- It can be a valuable alternative to consider for supplementing both mammalian and microbial processes
- Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Specifications
Specifications
| Certifications/Compliance | LUO |
| Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Format | Bottle |
| For Use With (Application) | Enzymatic Hydrolysate of Soybean meal |
| Product Line | Bacto |
| Quantity | 10 kg |
| Source | Animal origin, porcine |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.