Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Bacto™ TC Lactalbumin Hydrolysate
GREENER_CHOICE

Catalog No. 259961
Change view
Click to view available options
Quantity:
500 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-996-1 10 kg
DF5996-17-0 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-996-1 Supplier Gibco™ Supplier No. 259961
Add to Cart
Add to Cart

An animal origin peptone from the enzymatically hydrolyzed protein portion of milk whey.

This supplement is a mixture of peptides, amino acids, and carbohydrates, both simple and complex. Customers have successfully used Gibco™ Bacto™ TC Lactalbumin Hydrolysate to support the growth of many mammalian, bacterial, and insect cell types, and it has been used to produce both recombinant proteins and animal health vaccines. Customers have also used this supplement as an amino acid supplement for mammalian cell culture media.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Culture Type Mammalian Cell Culture
Form Powder
Product Type Dehydrated Culture Media
For Use With (Application) Mammalian, bacterial, insect cell types, recombinant proteins and animal health vaccines
Product Line Bacto
Quantity 10 kg
Source Animal Origin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.