Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Baculovirus Envelope gp64 Monoclonal Antibody (AcV1), PE, eBioscience™, Invitrogen™

Catalog No. 5011114
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-111-14 25 μg
50-111-15 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-111-14 Supplier Invitrogen™ Supplier No. 12699180
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The AcV1 monoclonal antibody reacts with the gp64 envelope protein of the baculovirus Autographa californica (AcMNPV). The gp64 envelope protein is essential for virus infectivity and is expressed on the surface of baculovirus-infected cells within six hours of infection. The AcV1 antibody can be used in flow cytometry-based viral titration experiments. Applications Reported: This AcV1 antibody has been reported for use in flow cytometric analysis. This antibody can be used in identifying virally-infected insect cells and determining viral titers. Applications Tested: This AcV1 antibody has been tested by flow cytometric analysis of baculovirus-infected insect cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Baculoviruses are known to infect invertebrates, with over 600 species having been described. Immature (larval) forms of moth species are the most common hosts, but these viruses have also been found infecting sawflies, mosquitoes, and shrimp. Although baculoviruses are capable of entering mammalian cells in culture, they are not known to be capable of replication in mammalian or other vertebrate animal cells. The gp64 envelope protein is essential for virus infectivity and is expressed on the surface of baculovirus-infected cells within six hours of infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Baculovirus Envelope gp64
Applications Flow Cytometry
Classification Monoclonal
Clone AcV1
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias BV gp64
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.