Learn More
Enzo Life Sciences Bad monoclonal antibody (5E6) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905638100
Encompass_Preferred
Clone: 5E6. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human Bad. UniProt ID: Q92934. Species reactivity: Human. Applications: ICC, IHC, IP, WB. Recommended Dilutions: Immunohistochemistry (1:50) Immunoprecipitation (1-2µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-200-6977
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.