Learn More
Enzo Life Sciences Bad polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADIAAP020D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of mouse Bad. Source: Purified from rabbit serum. Species reactivity: Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Drosophila, Bovine. Creossreactivity: Weakly cross-reacts with dog and chicken Bad. Applications: IHC, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7146
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.