Learn More
Leica Biosystems BADGE MONITOR XYLENE
Supplier:  Leica Biosystems 3801852
Encompass
Airchek Badges provide an innovative, simple and effective system to monitor vapour levels. Badges are available for formaldehyde, glutaraldehyde and xylene. The Airchek badges provide continuous sampling for personal monitoring and can give the measurements required under Government regulations. The badges are simply clipped on to the technician's clothing and a record is kept of name, location and time. One badge will provide continuous sampling over 8 hours and a second badge can be used to supply information to analyse a 15-minute period during peak exposure. 
Catalog No. NC0452335
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.