Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAF53A/BAF53B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518272
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518272 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518272 Supplier Invitrogen™ Supplier No. PA518272
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

The protein encoded by this gene is a member of a family of actin-related proteins which share significant amino acid sequence identity to conventional actins. Both actins and ARPs have an actin fold, which is an ATP-binding cleft, as a common feature. The ARPs are involved in diverse cellular processes, including vesicular transport, spindle orientation, nuclear migration and chromatin remodeling. This gene encodes a subunit of the BAF complex in mammals, which is functionally related to SWI/SNF complex in S. cerevisiae and Drosophila; the latter is thought to facilitate transcriptional activation of specific genes by antagonizing chromatin-mediated transcriptional repression. This subunit may be involved in the regulation of genes by structural modulation of their chromatin, specifically in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAF53A/BAF53B
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ACTL6B
Gene Accession No. O94805, O96019, P86173, Q99MR0, Q9Z2N8
Gene Alias 2810432C06Rik; 53 kDa BRG1-associated factor A; 53 kDa BRG1-associated factor B; actin like 6A; actin like 6B; actin-like 6; actin-like 6A; actin-like 6B; actin-like protein 6A; Actin-like protein 6B; actin-related protein 4; actin-related protein Baf53a; Actin-related protein Baf53b; Actl6; Actl6a; ACTL6B; AI851094; Arp4; ArpNa; arpNalpha; ArpN-alpha; ArpNbeta; ARPN-BETA; BAF complex 53 kDa subunit; BAF53; Baf53a; BAF53B; BRG1/brm-associated factor 53A; BRG1-associated factor 53A; BRG1-associated factor 53B; C79802; hArpN alpha; hArpN beta; INO80 complex subunit K; INO80K; MGC5382 Arp4
Gene Symbols ACTL6A, ACTL6B
Host Species Goat
Immunogen Synthetic peptide sequence (YEEGGKQCVERKCP) corresponding to the C-terminus amino acids of ACTL6A / ACTL6B.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288563, 361925, 51412, 56456, 83766, 86
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.