Learn More
AdipoGen BAFF aa134-285 Soluble h
SDP
Supplier:  AdipoGen AG40B00163010
Encompass_Preferred
Protein. Human BAFF aa 134-285 is fused at the N-terminus to a FLAG -tag. Natural cleaved form. Source E. coli. Endotoxin content <0.01EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Binds to human and mouse BAFF-R, TACI and BCMA. Relative binding affinity EC50 for BAFF-R human ~3ng/ml. Purity >95% SDS-PAGE. BAFF is mainly produced by innate immune cells such as neutrophils, monocytes, macrophages, dendritic cells, follicular dendritic cells. T cells, activated B cells, some malignant B cells and also non-lymphoid cells like astrocytes, synoviocytes and epithelial cells can also produce BAFF. BAFF binds three distinct receptors BAFF-R, TACI and BCMA expressed predominantly on B cells, although activated T cells also express BAFF-R. BAFF is a master regulator of peripheral B cell survival, and together with IL-6, promotes Ig class-switching and plasma cell differentiation. Besides its major role in B cell biology, BAFF co-stimulates activated T cells.
Catalog No. 50-256-2936
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.