Learn More
Enzo Life Sciences BAFF monoclonal antibody (Buffy-2) (100 µg)
SDP
Supplier:  Enzo Life Sciences ALX804131C100
Encompass_Preferred
Synonyms: B cell-activating factor, BLyS, TALL-1, THANK, zTNF4, TNFSF 13B/20, CD257. Clone: Buffy-2. Host: Rat. Isotype: IgM. Immunogen: Recombinant human soluble BAFF (aa 83-285) (Prod. No. ALX-522-025). UniProt ID: Q9Y275. Source: Purified from ascites. Species reactivity: Human. Applications: Flow Cytometry, IHC (FS), IHC (PS), WB. Purification: Purified by IgM purification HP column. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-210-0506
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.