Learn More
AdipoGen BAFF-R ms mAb 9B9 Biotin
SDP
Supplier:  AdipoGen AG20B0034BC100
Encompass_Preferred
Monoclonal Antibody. Recognizes mouse BAFF-R. Isotype Rat IgG2b. Clone 9B9. Applications FACS, FUNC. Liquid. In PBS containing 0.02% sodium azide. Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.
Catalog No. 50-256-3381
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.