Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse BAFF R (TNFRSF13C) ELISA Kit

Catalog No. 502550921
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-255-0921 5 x 96 Tests
EMTNFRSF13C 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-255-0921 Supplier Invitrogen™ Supplier No. EMTNFRSF13CX5
Add to Cart
Add to Cart

ELISA

The Mouse BAFF R (TNFRSF13C) ELISA quantitates Ms BAFF R in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms BAFF R. Principle of the method The Mouse BAFF R solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Tumor necrosis factor receptor superfamily member 13C (TNFRSF13C), also known as BAFFR, is a protein in humans is encoded by the TNFRSF13C gene. The BAFFR gene is mapped to chromosome 22q13. 1-q13. 31. It has got 184 amino acid transmembrane protein which is 56% identical to the mouse protein. B cell-activating factor (BAFF) enhances B-cell survival in vitro and is a regulator of the peripheral B-cell population. BAFF plays a crucial role in B cell development and can function through receptors other than BCMA.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9D8D0
Assay Range 28.67 to 8000 pg/mL
Assay Sensitivity 28 pg/mL
Calibration Range 28 pg/mL to 7000 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following mouse cytokines tested: BTC, C5a, CCL6, CD48, CD6, Chemerin, Clusterin, CXCL15, Cystatin C, DAN, DLL4, EDAR, Endocan, Fetuin A, H60, IL-33, IL-7 R alpha, Kremen-1, Limitin, Lipocalin-2, LOX-1, Marapsin, MBL-2, Meteorin, Nope, NOV, Osteoactivin, O x 40, Ligand, P-Cadherin, Periostin, PlGF-2, Progranulin, Prostasin, Renin 1, Testican 3, TIM-1, TRAIL, Tryptase epsilon
Sufficient For 5 x 96 Assays
Target Species Mouse
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 72049
Gene Symbol 2010006P15Rik,BAFF-R,Baffr,Bcmd,Bcmd-1,Bcmd1,Lvis22
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Immunology
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym 2010006P15Rik, BAFF-R, Baffr, Bcmd, Bcmd-1, Bcmd1, Lvis22
Target BAFF Receptor (CD268)
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.